The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 7 - 8 | 1049 | 95 |
NM_001320822.1 | NP_001307751.1 | 5 - 6 | 571 | 95 | |
NM_007368.3 | NP_031394.2 | 5 - 6 | 583 | 95 | |
XM_011534841.2 | XP_011533143.1 | 5 - 6 | 648 | 95 | |
XM_011534842.2 | XP_011533144.1 | 5 - 6 | 708 | 95 | |
XM_017020438.1 | XP_016875927.1 | 7 - 8 | 997 | 95 | |
XM_017020439.1 | XP_016875928.1 | 6 - 7 | 794 | 95 | |
GenBank mRNA | AK128094.1 | - | 7 - 8 | 1018 | 95 |
AK289746.1 | - | 5 - 6 | 489 | 95 | |
AK294913.1 | - | 5 - 6 | 556 | 95 | |
AK295861.1 | - | 4 - 5 | 417 | 95 | |
AK308880.1 | - | 2 - 3 | 645 | 95 | |
AK313795.1 | - | 5 - 6 | 517 | 95 | |
BC038456.1 | - | 5 - 6 | 580 | 95 | |
BC047242.1 | - | 5 - 6 | 571 | 95 | |
X89399.1 | - | 5 - 6 | 495 | 95 |