The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006149.3 | NP_006140.1 | 4 - 5 | 689 | 85 |
XM_011526974.1 | XP_011525276.1 | 3 - 4 | 928 | 85 | |
GenBank mRNA | AB006781.1 | - | 4 - 5 | 531 | 85 |
AF014838.1 | - | 4 - 5 | 488 | 85 | |
AK290269.1 | - | 4 - 5 | 534 | 85 | |
BC003661.2 | - | 4 - 5 | 518 | 85 | |
BC005146.2 | - | 4 - 5 | 518 | 85 | |
BC034750.1 | - | 4 - 5 | 514 | 85 | |
BT007032.1 | - | 4 - 5 | 475 | 85 | |
CR536544.1 | - | 4 - 5 | 475 | 85 | |
U82953.1 | - | 4 - 5 | 498 | 85 |