The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000384.2 | NP_000375.2 | 19 - 20 | 3123 | 71 |
GenBank mRNA | AK290844.1 | - | 19 - 20 | 3123 | 71 |
HM487065.1 | - | 19 - 20 | 2995 | 71 | |
J02610.1 | - | 19 - 20 | 3073 | 71 | |
M12480.1 | - | 9 - 10 | 1543 | 71 | |
M14081.1 | - | 19 - 20 | 2998 | 71 | |
M14162.1 | - | 19 - 20 | 2995 | 71 | |
X04506.1 | - | 19 - 20 | 3123 | 71 | |
X04714.1 | - | 19 - 20 | 3114 | 71 |