The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000384.2 | NP_000375.2 | 21 - 22 | 3452 | 124 |
GenBank mRNA | HM487065.1 | - | 21 - 22 | 3324 | 124 |
J02610.1 | - | 21 - 22 | 3402 | 124 | |
M14081.1 | - | 21 - 22 | 3327 | 124 | |
M14162.1 | - | 21 - 22 | 3324 | 124 | |
X04506.1 | - | 21 - 22 | 3452 | 124 |