The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000384.2 | NP_000375.2 | 4 - 5 | 505 | 143 |
GenBank mRNA | AK290844.1 | - | 4 - 5 | 505 | 143 |
BC051278.1 | - | 4 - 5 | 385 | 143 | |
HM487065.1 | - | 4 - 5 | 377 | 143 | |
J02610.1 | - | 4 - 5 | 455 | 143 | |
M12681.1 | - | 4 - 5 | 463 | 143 | |
M14081.1 | - | 4 - 5 | 377 | 143 | |
M14162.1 | - | 4 - 5 | 377 | 143 | |
X04506.1 | - | 4 - 5 | 505 | 143 | |
X04714.1 | - | 4 - 5 | 496 | 143 |