The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005113.3 | NP_005104.3 | 11 - 12 | 2309 | 113 |
GenBank mRNA | AF085199.1 | - | 11 - 12 | 2184 | 113 |
AK293077.1 | - | 11 - 12 | 2212 | 113 | |
AK303789.1 | - | 12 - 13 | 1942 | 113 | |
AY644768.1 | - | 10 - 11 | 2089 | 113 | |
BC023021.1 | - | 11 - 12 | 2217 | 113 |