The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005113.3 | NP_005104.3 | 5 - 6 | 1369 | 95 |
XM_011537420.2 | XP_011535722.1 | 5 - 6 | 1369 | 95 | |
GenBank mRNA | AF085199.1 | - | 5 - 6 | 1244 | 95 |
AJ132949.1 | - | 4 - 5 | 1113 | 95 | |
AK293077.1 | - | 5 - 6 | 1272 | 95 | |
AK303789.1 | - | 6 - 7 | 1002 | 95 | |
AY644768.1 | - | 4 - 5 | 1149 | 95 | |
BC023021.1 | - | 5 - 6 | 1277 | 95 |