Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK7
Entrez Gene ID
10089
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
K2p7.1, TWIK3
Chromosome Location
Chr.11: 65592855 - 65595996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005714.1 NP_005705.1 1 - 2 542 64
NM_033347.1 NP_203133.1 1 - 2 542 64
NM_033348.1 NP_203134.1 1 - 2 542 64
NM_033455.1 NP_258416.1 1 - 2 542 64
GenBank mRNA AF110522.1 - 1 - 2 542 64
AF110523.1 - 1 - 2 542 64
AF110524.1 - 1 - 2 542 64
BC103809.1 - 1 - 2 355 64
BC103810.2 - 1 - 2 354 64
BC103811.2 - 1 - 2 354 64

View Full Product Details