The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080387.4 | NP_525126.2 | 1 - 2 | 225 | 115 |
XM_011520632.2 | XP_011518934.1 | 2 - 3 | 194 | 115 | |
XM_011520633.2 | XP_011518935.1 | 1 - 2 | 220 | 115 | |
GenBank mRNA | AF411850.1 | - | 1 - 2 | 95 | 115 |
AK314071.1 | - | 1 - 2 | 212 | 115 | |
AY115592.1 | - | 1 - 2 | 216 | 115 | |
AY297446.1 | - | 1 - 2 | 224 | 115 | |
BC032313.1 | - | 1 - 2 | 225 | 115 | |
BI909298.1 | - | 1 - 2 | 173 | 115 |