The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206920.2 | NP_996803.2 | 12 - 13 | 1522 | 153 |
GenBank mRNA | AL137659.1 | - | 11 - 12 | 1819 | 153 |
AY358419.1 | - | 12 - 13 | 1522 | 153 |