Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
Chromosome Location
Chr.9: 136852367 - 136860799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206920.2 NP_996803.2 7 - 8 843 125
GenBank mRNA AL137659.1 - 7 - 8 1229 125
AY358419.1 - 7 - 8 843 125

View Full Product Details