The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000625.4 | NP_000616.3 | 13 - 14 | 1819 | 70 |
XM_011524859.2 | XP_011523161.1 | 13 - 14 | 1845 | 70 | |
XM_011524860.2 | XP_011523162.1 | 12 - 13 | 1638 | 70 | |
XM_011524861.1 | XP_011523163.1 | 13 - 14 | 1817 | 70 | |
XM_011524862.1 | XP_011523164.1 | 11 - 12 | 1416 | 70 | |
GenBank mRNA | AB022318.1 | - | 11 - 12 | 1438 | 70 |
AF068236.1 | - | 13 - 14 | 1789 | 70 | |
AK304252.1 | - | 13 - 14 | 1817 | 70 | |
BC130283.1 | - | 13 - 14 | 1739 | 70 | |
BC144126.1 | - | 13 - 14 | 1739 | 70 | |
D26525.1 | - | 12 - 13 | 1569 | 70 | |
L09210.1 | - | 13 - 14 | 1761 | 70 | |
L24553.1 | - | 12 - 13 | 1631 | 70 | |
U05810.1 | - | 13 - 14 | 1749 | 70 | |
U20141.1 | - | 12 - 13 | 1555 | 70 | |
U31511.1 | - | 13 - 14 | 1660 | 70 | |
X73029.1 | - | 13 - 14 | 1780 | 70 |