Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282529.1 NP_001269458.1 13 - 14 1767 126
NM_018449.3 NP_060919.3 17 - 18 2081 126
GenBank mRNA AB040924.1 - 5 - 6 883 126
AK025801.1 - 6 - 7 971 126
AK026088.1 - 9 - 10 1249 126
AK074884.1 - 4 - 5 440 126
AK294952.1 - 17 - 18 1999 126
AK299183.1 - 11 - 12 1489 126
AK302335.1 - 15 - 16 1902 126
AK302754.1 - 16 - 17 1949 126
AK303493.1 - 13 - 14 1705 126

View Full Product Details