The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282529.1 | NP_001269458.1 | 16 - 17 | 2171 | 135 |
NM_001282530.1 | NP_001269459.1 | 4 - 5 | 714 | 135 | |
NM_018449.3 | NP_060919.3 | 20 - 21 | 2485 | 135 | |
GenBank mRNA | AB040924.1 | - | 8 - 9 | 1287 | 135 |
AK025357.1 | - | 1 - 2 | 127 | 135 | |
AK025801.1 | - | 9 - 10 | 1375 | 135 | |
AK026088.1 | - | 12 - 13 | 1653 | 135 | |
AK057564.1 | - | 1 - 2 | 1040 | 135 | |
AK074884.1 | - | 7 - 8 | 844 | 135 | |
AK125935.1 | - | 3 - 4 | 1969 | 135 | |
AK294952.1 | - | 20 - 21 | 2403 | 135 | |
AK299183.1 | - | 14 - 15 | 1893 | 135 | |
AK302754.1 | - | 19 - 20 | 2353 | 135 | |
AK303493.1 | - | 16 - 17 | 2109 | 135 | |
BC037416.1 | - | 1 - 2 | 118 | 135 |