The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018449.3 | NP_060919.3 | 7 - 8 | 687 | 158 |
GenBank mRNA | AK294952.1 | - | 7 - 8 | 605 | 158 |
BE410205.1 | - | 4 - 5 | 375 | 158 | |
BF332025.1 | - | 4 - 5 | 158 |