The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021021.3 | NP_066301.1 | 2 - 3 | 1015 | 75 |
XM_011517239.1 | XP_011515541.1 | 2 - 3 | 992 | 75 | |
GenBank mRNA | AF028828.1 | - | 2 - 3 | 509 | 75 |
AK026095.1 | - | 4 - 5 | 1158 | 75 | |
AK292655.1 | - | 2 - 3 | 1089 | 75 | |
BC098573.1 | - | 2 - 3 | 975 | 75 |