The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022455.4 | NP_071900.2 | 22 - 23 | 6605 | 65 |
NM_172349.2 | NP_758859.1 | 23 - 24 | 5820 | 65 | |
GenBank mRNA | AF322907.1 | - | 18 - 19 | 6176 | 65 |
AF380302.1 | - | 23 - 24 | 5761 | 65 | |
AF395588.1 | - | 22 - 23 | 6512 | 65 | |
AK025916.1 | - | 4 - 5 | 559 | 65 | |
AK026066.1 | - | 11 - 12 | 1722 | 65 | |
AL832983.1 | - | 7 - 8 | 1094 | 65 | |
AY049721.1 | - | 21 - 22 | 6485 | 65 | |
BC144629.1 | - | 23 - 24 | 6050 | 65 | |
BC150628.1 | - | 22 - 23 | 5695 | 65 |