The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022455.4 | NP_071900.2 | 1 - 2 | 116 | 103 |
GenBank mRNA | BC021961.2 | - | 1 - 2 | 150 | 103 |