The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012113.2 | NP_036245.1 | 8 - 9 | 1232 | 64 |
XM_006711261.2 | XP_006711324.1 | 8 - 9 | 1601 | 64 | |
XM_011509379.2 | XP_011507681.1 | 8 - 9 | 1678 | 64 | |
XM_017000894.1 | XP_016856383.1 | 6 - 7 | 936 | 64 | |
XM_017000896.1 | XP_016856385.1 | 6 - 7 | 1376 | 64 | |
GenBank mRNA | AB025904.1 | - | 8 - 9 | 1155 | 64 |
AK074765.1 | - | 8 - 9 | 1201 | 64 | |
AK290609.1 | - | 8 - 9 | 1071 | 64 | |
AK295341.1 | - | 6 - 7 | 1328 | 64 | |
AK315469.1 | - | 8 - 9 | 1164 | 64 | |
AY358689.1 | - | 8 - 9 | 1116 | 64 | |
BC034412.1 | - | 8 - 9 | 1086 | 64 | |
BT020054.1 | - | 8 - 9 | 846 | 64 |