The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012113.2 | NP_036245.1 | 1 - 2 | 440 | 65 |
XM_005245059.2 | XP_005245116.1 | 1 - 2 | 369 | 65 | |
XM_005245060.2 | XP_005245117.1 | 1 - 2 | 369 | 65 | |
XM_006711259.2 | XP_006711322.1 | 1 - 2 | 369 | 65 | |
XM_017000894.1 | XP_016856383.1 | 1 - 2 | 369 | 65 | |
XM_017000895.1 | XP_016856384.1 | 1 - 2 | 369 | 65 | |
GenBank mRNA | AB025904.1 | - | 1 - 2 | 363 | 65 |
AK074765.1 | - | 1 - 2 | 409 | 65 | |
AK290609.1 | - | 1 - 2 | 279 | 65 | |
AK315469.1 | - | 1 - 2 | 372 | 65 | |
AY358689.1 | - | 1 - 2 | 324 | 65 | |
BC034412.1 | - | 1 - 2 | 294 | 65 | |
BP361793.1 | - | 1 - 2 | 440 | 65 | |
BT020054.1 | - | 1 - 2 | 54 | 65 |