The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 16 - 17 | 2390 | 78 |
NM_013241.2 | NP_037373.2 | 14 - 15 | 2312 | 78 | |
XM_006721180.1 | XP_006721243.1 | 5 - 6 | 965 | 78 | |
XM_011523043.2 | XP_011521345.1 | 16 - 17 | 2606 | 78 | |
XM_011523044.1 | XP_011521346.1 | 16 - 17 | 2335 | 78 | |
XM_011523045.2 | XP_011521347.1 | 16 - 17 | 2100 | 78 | |
GenBank mRNA | AB041046.1 | - | 14 - 15 | 2282 | 78 |
AB209584.1 | - | 12 - 13 | 2781 | 78 | |
AF113615.1 | - | 14 - 15 | 2218 | 78 | |
AK301157.1 | - | 4 - 5 | 1023 | 78 | |
AY192154.1 | - | 14 - 15 | 2200 | 78 | |
BC033084.1 | - | 14 - 15 | 2257 | 78 | |
KJ658417.1 | - | 16 - 17 | 2278 | 78 |