Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
Chromosome Location
Chr.16: 67229389 - 67247522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318202.1 NP_001305131.1 20 - 21 3048 88
NM_013241.2 NP_037373.2 18 - 19 2970 88
XM_006721180.1 XP_006721243.1 9 - 10 1623 88
XM_011523043.2 XP_011521345.1 20 - 21 3264 88
XM_011523044.1 XP_011521346.1 19 - 20 2831 88
XM_011523045.2 XP_011521347.1 20 - 21 2758 88
GenBank mRNA AB041046.1 - 18 - 19 2940 88
AB209584.1 - 16 - 17 3439 88
AF113615.1 - 18 - 19 2876 88
AK025992.1 - 4 - 5 660 88
AK301157.1 - 8 - 9 1681 88
AY192154.1 - 18 - 19 2858 88
BC033084.1 - 18 - 19 2915 88
KJ658417.1 - 20 - 21 2936 88

View Full Product Details