The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 22 - 23 | 3409 | 71 |
NM_013241.2 | NP_037373.2 | 20 - 21 | 3331 | 71 | |
XM_006721180.1 | XP_006721243.1 | 11 - 12 | 1984 | 71 | |
XM_011523043.2 | XP_011521345.1 | 22 - 23 | 3625 | 71 | |
XM_011523044.1 | XP_011521346.1 | 21 - 22 | 3192 | 71 | |
XM_011523045.2 | XP_011521347.1 | 22 - 23 | 3119 | 71 | |
GenBank mRNA | AB041046.1 | - | 20 - 21 | 3301 | 71 |
AB209584.1 | - | 18 - 19 | 3800 | 71 | |
AF113615.1 | - | 20 - 21 | 3237 | 71 | |
AK025992.1 | - | 6 - 7 | 1021 | 71 | |
AK301157.1 | - | 10 - 11 | 2042 | 71 | |
AY192154.1 | - | 20 - 21 | 3219 | 71 | |
BC033084.1 | - | 20 - 21 | 3276 | 71 | |
BU689676.1 | - | 2 - 3 | 71 | ||
KJ658417.1 | - | 22 - 23 | 3297 | 71 |