Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX3
Entrez Gene ID
1747
Gene Name
distal-less homeobox 3
Gene Aliases
AI4, TDO
Chromosome Location
Chr.17: 49990005 - 49995224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005220.2 NP_005211.1 2 - 3 744 132
GenBank mRNA AK075167.1 - 2 - 3 734 132
AK313257.1 - 2 - 3 744 132
BC012361.1 - 2 - 3 723 132
BC028970.1 - 2 - 3 730 132

View Full Product Details