The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133459.3 | NP_597716.1 | 8 - 9 | 987 | 150 |
XM_017025556.1 | XP_016881045.1 | 8 - 9 | 1867 | 150 | |
XM_017025557.1 | XP_016881046.1 | 9 - 10 | 1760 | 150 | |
XM_017025558.1 | XP_016881047.1 | 8 - 9 | 1755 | 220 | |
GenBank mRNA | AB075863.1 | - | 8 - 9 | 955 | 150 |
AY358347.1 | - | 8 - 9 | 933 | 150 | |
BC046645.1 | - | 3 - 4 | 622 | 150 |