Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NSD1
- Entrez Gene ID
-
64324
- Gene Name
- nuclear receptor binding SET domain protein 1
- Gene Aliases
-
ARA267, KMT3B, SOTOS, SOTOS1, STO
- Chromosome Location
-
Chr.5: 177133079 - 177300213 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF322907.1 |
- |
1 - 2 |
952 |
126 |
View Full Product Details