The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024052.4 | NP_076957.3 | 3 - 4 | 772 | 120 |
XM_005256800.1 | XP_005256857.1 | 3 - 4 | 659 | 120 | |
XM_011524008.1 | XP_011522310.1 | 3 - 4 | 659 | 120 | |
GenBank mRNA | AK074284.1 | - | 3 - 4 | 647 | 120 |
AK127580.1 | - | 3 - 4 | 459 | 120 | |
AK315083.1 | - | 3 - 4 | 660 | 120 | |
BC000636.2 | - | 3 - 4 | 433 | 120 | |
BC041829.1 | - | 3 - 4 | 777 | 120 |