The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142293.1 | NP_001135765.1 | 5 - 6 | 649 | 68 |
NM_014570.4 | NP_055385.3 | 6 - 7 | 781 | 68 | |
XM_005261525.3 | XP_005261582.1 | 6 - 7 | 671 | 68 | |
GenBank mRNA | AF111847.1 | - | 6 - 7 | 618 | 68 |
AK002083.1 | - | 3 - 4 | 210 | 68 | |
AK022768.1 | - | 1 - 2 | 46 | 68 | |
AK297635.1 | - | 4 - 5 | 431 | 68 | |
AK300531.1 | - | 5 - 6 | 422 | 68 | |
AK307967.1 | - | 5 - 6 | 487 | 68 | |
AK315720.1 | - | 6 - 7 | 620 | 68 | |
AL137598.1 | - | 5 - 6 | 433 | 68 | |
AL159143.1 | - | 6 - 7 | 627 | 68 | |
BC005122.2 | - | 6 - 7 | 590 | 68 | |
CR456382.1 | - | 6 - 7 | 572 | 68 |