The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256371.1 | NP_001243300.1 | 11 - 12 | 2349 | 134 |
NM_001256372.1 | NP_001243301.1 | 11 - 12 | 2294 | 134 | |
NM_024680.3 | NP_078956.2 | 11 - 12 | 2548 | 134 | |
XM_011520367.1 | XP_011518669.1 | 10 - 11 | 2235 | 134 | |
GenBank mRNA | AK026964.1 | - | 6 - 7 | 1118 | 134 |
AK055206.1 | - | 8 - 9 | 1589 | 134 | |
AK225338.1 | - | 6 - 7 | 1124 | 134 | |
AK292688.1 | - | 11 - 12 | 2259 | 134 | |
BC028244.1 | - | 4 - 5 | 949 | 134 | |
BC090877.1 | - | 6 - 7 | 1151 | 134 | |
BC126400.1 | - | 11 - 12 | 2539 | 134 | |
BC126402.1 | - | 11 - 12 | 2539 | 134 |