Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPM3
Entrez Gene ID
7170
Gene Name
tropomyosin 3
Gene Aliases
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
Chromosome Location
Chr.1: 154155304 - 154192135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278189.1 NP_001265118.1 7 - 8 792 75
NM_152263.3 NP_689476.2 8 - 9 890 75
GenBank mRNA AF474157.1 - 7 - 8 662 75
AK056997.1 - 8 - 9 680 75
BC008407.1 - 8 - 9 856 75
BC008425.1 - 8 - 9 859 75
BC062740.1 - 8 - 9 722 75
BM674269.1 - 2 - 3 75
CR997359.1 - 7 - 8 683 75
KU178369.1 - 7 - 8 662 75
X04201.1 - 8 - 9 831 75

View Full Product Details