Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TPM3
- Entrez Gene ID
-
7170
- Gene Name
- tropomyosin 3
- Gene Aliases
-
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
- Chromosome Location
-
Chr.1: 154155304 - 154192135 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BM674269.1 |
- |
1 - 2 |
451 |
63 |
View Full Product Details