The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110798.1 | NP_001104268.1 | 4 - 5 | 730 | 84 |
NM_018527.3 | NP_060997.2 | 4 - 5 | 730 | 84 | |
NM_024561.4 | NP_078837.3 | 4 - 5 | 730 | 84 | |
XM_006719866.2 | XP_006719929.1 | 4 - 5 | 706 | 84 | |
XM_011535227.2 | XP_011533529.1 | 4 - 5 | 706 | 84 | |
XM_011535228.1 | XP_011533530.1 | 4 - 5 | 706 | 84 | |
XM_017020744.1 | XP_016876233.1 | 1 - 2 | 122 | 84 | |
XM_017020745.1 | XP_016876234.1 | 4 - 5 | 706 | 84 | |
XM_017020746.1 | XP_016876235.1 | 4 - 5 | 706 | 84 | |
GenBank mRNA | AK097224.1 | - | 4 - 5 | 640 | 84 |
AK291248.1 | - | 4 - 5 | 644 | 84 | |
BC032318.1 | - | 4 - 5 | 653 | 84 | |
BC064592.1 | - | 4 - 5 | 655 | 84 | |
BC101719.1 | - | 4 - 5 | 502 | 84 | |
BC101721.1 | - | 4 - 5 | 502 | 84 | |
BX640665.1 | - | 4 - 5 | 624 | 84 |