Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA16
Entrez Gene ID
79612
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases
NARG1L
Chromosome Location
Chr.13: 41311205 - 41377031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018527.3 NP_060997.2 8 - 9 1225 101
XM_005266523.2 XP_005266580.1 2 - 3 184 101
GenBank mRNA BC032318.1 - 8 - 9 1148 101

View Full Product Details