The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015130.2 | NP_055945.2 | 7 - 8 | 1537 | 98 |
GenBank mRNA | AB020689.2 | - | 7 - 8 | 1340 | 98 |
AB449892.1 | - | 7 - 8 | 1265 | 98 | |
AK127020.1 | - | 4 - 5 | 825 | 98 | |
BC146758.1 | - | 7 - 8 | 1340 | 98 | |
CN332501.1 | - | 2 - 3 | 284 | 98 |