Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZZZ3
Entrez Gene ID
26009
Gene Name
zinc finger ZZ-type containing 3
Gene Aliases
ATAC1
Chromosome Location
Chr.1: 77561526 - 77683440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015534.5 NP_056349.1 5 - 6 1973 124
XM_005270725.3 XP_005270782.1 5 - 6 2048 124
XM_005270726.3 XP_005270783.1 4 - 5 1949 124
XM_005270727.3 XP_005270784.1 5 - 6 2048 124
XM_017000970.1 XP_016856459.1 4 - 5 1949 124
GenBank mRNA AK074119.1 - 1 - 2 1019 124
BC035397.1 - 1 - 2 644 124
BC035818.2 - 1 - 2 1548 124
BX640766.1 - 1 - 2 1250 124
BX641001.1 - 5 - 6 1974 124
BX641096.1 - 5 - 6 1959 124

View Full Product Details