The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271878.1 | NP_001258807.1 | 2 - 3 | 807 | 92 |
GenBank mRNA | U78971.1 | - | 2 - 3 | 505 | 92 |