The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000919.3 | NP_000910.2 | 22 - 23 | 2854 | 118 |
NM_001177306.1 | NP_001170777.1 | 22 - 23 | 2854 | 118 | |
NM_001319943.1 | NP_001306872.1 | 24 - 25 | 3474 | 118 | |
NM_138821.2 | NP_620176.1 | 21 - 22 | 2533 | 118 | |
XM_011543419.2 | XP_011541721.1 | 24 - 25 | 3028 | 118 | |
XM_017009497.1 | XP_016864986.1 | 23 - 24 | 2974 | 118 | |
XM_017009498.1 | XP_016864987.1 | 23 - 24 | 2904 | 118 | |
XM_017009501.1 | XP_016864990.1 | 22 - 23 | 2850 | 118 | |
XM_017009507.1 | XP_016864996.1 | 22 - 23 | 2653 | 118 | |
XM_017009510.1 | XP_016864999.1 | 21 - 22 | 2633 | 118 | |
GenBank mRNA | AB095007.1 | - | 22 - 23 | 2481 | 118 |
AF035320.1 | - | 4 - 5 | 365 | 118 | |
AK290158.1 | - | 22 - 23 | 2592 | 118 | |
BC018127.1 | - | 21 - 22 | 2533 | 118 | |
BT007419.1 | - | 21 - 22 | 2160 | 118 | |
M37721.1 | - | 22 - 23 | 2669 | 118 |