Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAM
Entrez Gene ID
5066
Gene Name
peptidylglycine alpha-amidating monooxygenase
Gene Aliases
PAL, PHM
Chromosome Location
Chr.5: 102754783 - 103031105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000919.3 NP_000910.2 7 - 8 947 73
NM_001177306.1 NP_001170777.1 7 - 8 947 73
NM_001319943.1 NP_001306872.1 8 - 9 1513 73
NM_138766.2 NP_620121.1 7 - 8 947 73
NM_138821.2 NP_620176.1 7 - 8 947 73
NM_138822.2 NP_620177.1 7 - 8 947 73
NR_033440.1 6 - 7 889 73
XM_011543419.2 XP_011541721.1 8 - 9 1067 73
XM_017009495.1 XP_016864984.1 7 - 8 943 73
XM_017009496.1 XP_016864985.1 7 - 8 943 73
XM_017009497.1 XP_016864986.1 8 - 9 1067 73
XM_017009498.1 XP_016864987.1 7 - 8 943 73
XM_017009499.1 XP_016864988.1 7 - 8 943 73
XM_017009500.1 XP_016864989.1 7 - 8 943 73
XM_017009501.1 XP_016864990.1 7 - 8 943 73
XM_017009502.1 XP_016864991.1 7 - 8 943 73
XM_017009503.1 XP_016864992.1 7 - 8 943 73
XM_017009504.1 XP_016864993.1 7 - 8 1044 73
XM_017009505.1 XP_016864994.1 8 - 9 1067 73
XM_017009506.1 XP_016864995.1 7 - 8 943 73
XM_017009507.1 XP_016864996.1 8 - 9 1067 73
XM_017009508.1 XP_016864997.1 7 - 8 943 73
XM_017009509.1 XP_016864998.1 7 - 8 943 73
XM_017009510.1 XP_016864999.1 7 - 8 1047 73
XM_017009511.1 XP_016865000.1 7 - 8 943 73
XM_017009512.1 XP_016865001.1 7 - 8 943 73
XM_017009513.1 XP_016865002.1 7 - 8 943 73
XM_017009514.1 XP_016865003.1 7 - 8 943 73
XM_017009515.1 XP_016865004.1 7 - 8 943 73
XM_017009516.1 XP_016865005.1 7 - 8 943 73
GenBank mRNA AB095007.1 - 7 - 8 574 73
AF010472.1 - 6 - 7 490 73
AK290158.1 - 7 - 8 685 73
AK296521.1 - 6 - 7 655 73
BC018127.1 - 7 - 8 947 73
BT007419.1 - 7 - 8 574 73
M37721.1 - 7 - 8 762 73
S75037.1 - 7 - 8 654 73
S75038.1 - 7 - 8 654 73

View Full Product Details