Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
Chromosome Location
Chr.16: 67229389 - 67247522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721180.1 XP_006721243.1 2 - 3 86 67

View Full Product Details