Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
Chromosome Location
Chr.2: 232697305 - 232860577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103146.1 NP_001096616.1 28 - 29 4023 86
NM_001103147.1 NP_001096617.1 30 - 31 4156 86
NM_001103148.1 NP_001096618.1 27 - 28 3939 86
NM_015575.3 NP_056390.2 30 - 31 4163 86
NR_103492.1 26 - 27 3784 86
GenBank mRNA AB014542.2 - 30 - 31 4115 86
AK027021.1 - 7 - 8 1305 86
AY176045.1 - 30 - 31 4137 86
BC008072.2 - 4 - 5 664 86
BC136251.1 - 27 - 28 3891 86
BC143352.1 - 26 - 27 3736 86
BC146775.1 - 30 - 31 4115 86
BX537885.1 - 28 - 29 3955 86
BX538172.1 - 28 - 29 3886 86
BX538321.1 - 28 - 29 3979 86
BX647172.1 - 12 - 13 2858 86

View Full Product Details