The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103146.1 | NP_001096616.1 | 8 - 9 | 730 | 92 |
NM_001103148.1 | NP_001096618.1 | 7 - 8 | 664 | 92 | |
NM_015575.3 | NP_056390.2 | 10 - 11 | 870 | 92 | |
NR_103492.1 | 7 - 8 | 646 | 92 | ||
GenBank mRNA | AK001739.1 | - | 10 - 11 | 844 | 92 |
AK299418.1 | - | 9 - 10 | 774 | 92 | |
AY176045.1 | - | 10 - 11 | 844 | 92 | |
BC043402.1 | - | 8 - 9 | 683 | 92 | |
BC136251.1 | - | 7 - 8 | 601 | 92 | |
BC143352.1 | - | 7 - 8 | 601 | 92 | |
BX537885.1 | - | 8 - 9 | 665 | 92 | |
BX538172.1 | - | 7 - 8 | 632 | 92 | |
BX538321.1 | - | 8 - 9 | 686 | 92 | |
BX647505.1 | - | 7 - 8 | 477 | 92 |