The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006080.2 | NP_006071.1 | 11 - 12 | 1677 | 124 |
XM_005250110.3 | XP_005250167.1 | 14 - 15 | 1872 | 124 | |
XM_005250111.4 | XP_005250168.1 | 13 - 14 | 1712 | 124 | |
XM_006715839.3 | XP_006715902.1 | 12 - 13 | 1615 | 124 | |
XM_011515734.2 | XP_011514036.1 | 14 - 15 | 1697 | 124 | |
XM_017011673.1 | XP_016867162.1 | 12 - 13 | 1532 | 124 | |
GenBank mRNA | AK289954.1 | - | 11 - 12 | 1770 | 124 |
BC111416.1 | - | 11 - 12 | 1376 | 124 | |
L26081.1 | - | 11 - 12 | 1377 | 124 |