Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA3A
Entrez Gene ID
10371
Gene Name
semaphorin 3A
Gene Aliases
COLL1, HH16, Hsema-I, Hsema-III, SEMA1, SEMAD, SEMAIII, SEMAL, SemD, coll-1
Chromosome Location
Chr.7: 83956846 - 84492724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006080.2 NP_006071.1 3 - 4 645 93
XM_005250110.3 XP_005250167.1 6 - 7 840 93
XM_005250111.4 XP_005250168.1 5 - 6 680 93
XM_006715839.3 XP_006715902.1 4 - 5 583 93
XM_011515734.2 XP_011514036.1 6 - 7 665 93
XM_017011673.1 XP_016867162.1 4 - 5 500 93
GenBank mRNA AK289954.1 - 3 - 4 738 93
BC111416.1 - 3 - 4 344 93
DR002146.1 - 3 - 4 483 93
L26081.1 - 3 - 4 345 93

View Full Product Details