Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
Chromosome Location
Chr.1: 27355179 - 27368037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297609.1 NP_001284538.1 19 - 20 3023 73
NM_004672.4 NP_004663.3 20 - 21 3047 73
XM_017002771.1 XP_016858260.1 18 - 19 2428 73
GenBank mRNA AB167411.1 - 20 - 21 2778 73
AB208805.1 - 19 - 20 2492 73
AF100318.1 - 17 - 18 2192 73
BC015914.1 - 16 - 17 1949 73
BC109032.2 - 11 - 12 1326 73
BC129950.1 - 12 - 13 1477 73
BC129951.1 - 12 - 13 1477 73

View Full Product Details