The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297609.1 | NP_001284538.1 | 26 - 27 | 3956 | 73 |
NM_004672.4 | NP_004663.3 | 27 - 28 | 3980 | 73 | |
XM_017002771.1 | XP_016858260.1 | 25 - 26 | 3361 | 73 | |
GenBank mRNA | AB167411.1 | - | 27 - 28 | 3711 | 73 |
AB208805.1 | - | 26 - 27 | 3425 | 73 | |
AF100318.1 | - | 24 - 25 | 3125 | 73 | |
BC129950.1 | - | 19 - 20 | 2410 | 73 | |
BC129951.1 | - | 19 - 20 | 2410 | 73 |