The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297609.1 | NP_001284538.1 | 5 - 6 | 1219 | 102 |
NM_004672.4 | NP_004663.3 | 6 - 7 | 1243 | 102 | |
XM_017002771.1 | XP_016858260.1 | 4 - 5 | 612 | 102 | |
XM_017002772.1 | XP_016858261.1 | 6 - 7 | 2414 | 102 | |
GenBank mRNA | AB167411.1 | - | 6 - 7 | 974 | 102 |
AB208805.1 | - | 5 - 6 | 688 | 102 | |
AF100318.1 | - | 3 - 4 | 388 | 102 | |
BC015914.1 | - | 2 - 3 | 145 | 102 |