Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF4
Entrez Gene ID
26094
Gene Name
DDB1 and CUL4 associated factor 4
Gene Aliases
WDR21, WDR21A
Chromosome Location
Chr.14: 72926332 - 72962098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163508.1 NP_001156980.1 4 - 5 577 92
NM_001163509.1 NP_001156981.1 4 - 5 545 92
NM_015604.3 NP_056419.2 4 - 5 577 92
NM_181340.2 NP_851937.1 3 - 4 477 92
XM_017021201.1 XP_016876690.1 3 - 4 386 92
XM_017021202.1 XP_016876691.1 3 - 4 386 92
XM_017021203.1 XP_016876692.1 4 - 5 572 92
XM_017021204.1 XP_016876693.1 4 - 5 542 92
XM_017021205.1 XP_016876694.1 5 - 6 711 92
XM_017021206.1 XP_016876695.1 4 - 5 547 92
XM_017021207.1 XP_016876696.1 4 - 5 572 92
XM_017021208.1 XP_016876697.1 4 - 5 572 92
XM_017021209.1 XP_016876698.1 4 - 5 572 92
XM_017021210.1 XP_016876699.1 4 - 5 572 92
XM_017021211.1 XP_016876700.1 4 - 5 516 92
XM_017021212.1 XP_016876701.1 5 - 6 679 92
XM_017021213.1 XP_016876702.1 4 - 5 479 92
XM_017021214.1 XP_016876703.1 4 - 5 528 92
XM_017021215.1 XP_016876704.1 4 - 5 546 92
GenBank mRNA AK027745.1 - 4 - 5 422 92
AK297571.1 - 4 - 5 426 92
AK297742.1 - 4 - 5 388 92
AK315499.1 - 4 - 5 577 92
AL080157.1 - 3 - 4 348 92
BC018979.2 - 4 - 5 389 92
BC035901.1 - 3 - 4 331 92
BX247962.1 - 4 - 5 404 92

View Full Product Details