The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198842.1 | NP_001185771.1 | 5 - 6 | 664 | 90 |
NM_006431.2 | NP_006422.1 | 5 - 6 | 433 | 90 | |
GenBank mRNA | AB451223.1 | - | 4 - 5 | 339 | 90 |
AB451346.1 | - | 4 - 5 | 339 | 90 | |
AF026166.1 | - | 5 - 6 | 396 | 90 | |
AF026293.1 | - | 5 - 6 | 372 | 90 | |
AK290767.1 | - | 5 - 6 | 428 | 90 | |
AK294307.1 | - | 5 - 6 | 678 | 90 | |
AK302157.1 | - | 5 - 6 | 90 | ||
AK316397.1 | - | 5 - 6 | 603 | 90 | |
AK316408.1 | - | 5 - 6 | 664 | 90 | |
BC113514.1 | - | 5 - 6 | 411 | 90 | |
BC113516.1 | - | 5 - 6 | 411 | 90 | |
BT019966.1 | - | 4 - 5 | 339 | 90 | |
FJ224351.1 | - | 5 - 6 | 372 | 90 |