The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198842.1 | NP_001185771.1 | 6 - 7 | 769 | 92 |
NM_006431.2 | NP_006422.1 | 6 - 7 | 538 | 92 | |
GenBank mRNA | AB451223.1 | - | 5 - 6 | 444 | 92 |
AB451346.1 | - | 5 - 6 | 444 | 92 | |
AF026166.1 | - | 6 - 7 | 501 | 92 | |
AF026293.1 | - | 6 - 7 | 477 | 92 | |
AK290767.1 | - | 6 - 7 | 533 | 92 | |
AK294307.1 | - | 6 - 7 | 783 | 92 | |
AK302157.1 | - | 6 - 7 | 578 | 92 | |
AK316397.1 | - | 6 - 7 | 708 | 92 | |
AK316408.1 | - | 6 - 7 | 769 | 92 | |
BC113514.1 | - | 6 - 7 | 516 | 92 | |
BC113516.1 | - | 6 - 7 | 516 | 92 | |
BT019966.1 | - | 5 - 6 | 444 | 92 | |
CR457071.1 | - | 5 - 6 | 444 | 92 | |
FJ224351.1 | - | 6 - 7 | 477 | 92 |