The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198842.1 | NP_001185771.1 | 8 - 9 | 1078 | 94 |
NM_006431.2 | NP_006422.1 | 8 - 9 | 847 | 94 | |
GenBank mRNA | AB451223.1 | - | 7 - 8 | 753 | 94 |
AB451346.1 | - | 7 - 8 | 753 | 94 | |
AF026166.1 | - | 8 - 9 | 810 | 94 | |
AF026293.1 | - | 8 - 9 | 786 | 94 | |
AK290767.1 | - | 8 - 9 | 842 | 94 | |
AK294307.1 | - | 8 - 9 | 1092 | 94 | |
AK302157.1 | - | 8 - 9 | 887 | 94 | |
AK316397.1 | - | 8 - 9 | 1017 | 94 | |
AK316408.1 | - | 8 - 9 | 1078 | 94 | |
BC113514.1 | - | 8 - 9 | 825 | 94 | |
BC113516.1 | - | 8 - 9 | 825 | 94 | |
BT019966.1 | - | 7 - 8 | 753 | 94 | |
CR457071.1 | - | 7 - 8 | 753 | 94 | |
FJ224351.1 | - | 8 - 9 | 786 | 94 |